Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fuom | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106310.1 | NP_001099780.1 | 5 | 780 | ||
GenBank mRNA | FQ212771.1 | - | 4 | 508 |
Gene Symbol: LOC100911225 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748887.3 | XP_003748935.1 | 5 | 402 |