Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Vipr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017238.1 | NP_058934.1 | 5 | 545 | ||
GenBank mRNA | U09631.1 | - | 5 | 545 | ||
Z25885.1 | - | 5 | 498 |
Set Membership: |
![]() |