Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Arpc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106615.2 | NP_001100085.2 | 5 | 481 | ||
GenBank mRNA | BC167049.1 | - | 5 | 469 | 7442696 | |
CK478610.1 | - | 5 | 481 | |||
FQ228740.1 | - | 5 | 420 |