Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pole2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001169108.1 | NP_001162579.1 | 7 | 549 | ||
XM_006240154.2 | XP_006240216.1 | 7 | 564 | |||
XM_008764696.1 | XP_008762918.1 | 10 | 1348 | |||
XM_008764697.1 | XP_008762919.1 | 9 | 1158 | |||
XM_008764698.1 | XP_008762920.1 | 10 | 1287 | |||
XM_008764699.1 | XP_008762921.1 | 10 | 1371 | |||
XM_008764700.1 | XP_008762922.1 | 10 | 1257 | |||
XM_008764701.1 | XP_008762923.1 | 10 | 1151 | |||
XM_008764702.1 | XP_008762924.1 | 10 | 1247 |