Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tbx22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001106937.1 | NP_001100407.1 | 6, 7 | 838 | ||
XM_006257159.2 | XP_006257221.1 | 7, 8 | 1107 | |||
XM_006257160.2 | XP_006257222.1 | 7, 8 | 957 | |||
XM_006257161.2 | XP_006257223.1 | 7, 8 | 1104 | |||
XM_006257162.2 | XP_006257224.1 | 6, 7 | 1147 | |||
XM_008773339.1 | XP_008771561.1 | 6, 7 | 822 |