Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Acat2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006995.1 | NP_001006996.1 | 8 | 974 | ||
GenBank mRNA | AY325187.1 | - | 948 | |||
BC083872.1 | - | 8 | 974 | 7128564 |