Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Slc30a7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001191715.1 | NP_001178644.1 | 7 | 850 | ||
GenBank mRNA | BC091417.1 | - | 3 | 208 | ||
BC107441.1 | - | 3 | 209 |