Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Clec4a3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005891.1 | NP_001005891.1 | 3 | 306 | ||
XM_006237321.2 | XP_006237383.1 | 3 | 515 | |||
GenBank mRNA | AY397671.1 | - | 3 | 306 | ||
AY494062.1 | - | 3 | 307 | |||
FQ230051.1 | - | 3 | 401 | |||
FQ234061.1 | - | 3 | 431 |