Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000539.1 | NP_001000539.1 | 1 | 471 |
Gene Symbol: LOC100910456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003748751.2 | XP_003748799.2 | 1 | 195 |
Set Membership: |
G-protein coupled receptor |