Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Upk1a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001108911.1 | NP_001102381.1 | 4 | 497 | ||
XM_006228782.2 | XP_006228844.1 | 6 | 854 | |||
XM_006228784.2 | XP_006228846.1 | 6 | 854 | |||
XM_008759176.1 | XP_008757398.1 | 5 | 834 |