Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Prl3d1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006253914.2 | 3 | 274 | |||
GenBank mRNA | FQ220053.1 | - | 278 | |||
FQ220072.1 | - | 274 | ||||
FQ220274.1 | - | 276 | ||||
FQ220568.1 | - | 276 | ||||
FQ220587.1 | - | 274 | ||||
FQ221231.1 | - | 276 | ||||
FQ222593.1 | - | 274 | ||||
FQ223300.1 | - | 277 | ||||
FQ228787.1 | - | 274 | ||||
FQ229328.1 | - | 274 | ||||
FQ229656.1 | - | 268 | ||||
FQ230119.1 | - | 274 | ||||
M55269.1 | - | 267 |
Gene Symbol: LOC103689989 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771673.1 | XP_008769895.1 | 216 | |||
XM_008771674.1 | XP_008769896.1 | 254 |
Gene Symbol: LOC103690150 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771672.1 | XP_008769894.1 | 251 | |||
GenBank mRNA | FQ228618.1 | - | 274 |
Gene Symbol: Prl3d2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771671.1 | XP_008769893.1 | 3 | 277 | ||
GenBank mRNA | BC161851.1 | - | 3 | 251 | 9088452 | |
D21103.1 | - | 3 | 213 | |||
DQ329282.1 | - | 3 | 270 | |||
DQ329283.1 | - | 3 | 270 | |||
FQ145043.1 | - | 3 | 276 | |||
FQ221365.1 | - | 3 | 274 | |||
FQ223255.1 | - | 3 | 277 | |||
FQ223518.1 | - | 3 | 274 | |||
FQ229928.1 | - | 3 | 276 | |||
FQ230035.1 | - | 3 | 274 |
Gene Symbol: Prl3d4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033233.1 | NP_150236.1 | 3 | 234 | ||
XM_008771595.1 | XP_008769817.1 | 3 | 216 | |||
GenBank mRNA | BC097302.1 | - | 3 | 260 | 7444366 | |
U32679.1 | - | 3 | 234 |