Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Eif2ak2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019335.1 | NP_062208.1 | 12 | 1133 | ||
XM_008764425.1 | XP_008762647.1 | 11 | 1357 | |||
XM_008764426.1 | XP_008762648.1 | 12 | 1199 | |||
GenBank mRNA | L29281.1 | - | 12 | 1133 |