Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NXPH2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_007226.2 | NP_009157.1 | 2 | 715 | ||
GenBank mRNA | AB162675.1 | - | 2 | 715 | ||
AF043467.1 | - | 2 | 709 | |||
BC101461.1 | - | 2 | 715 | 40022686 | ||
BC101462.1 | - | 2 | 715 | 40022687 | ||
BC101463.1 | - | 2 | 715 | 40022688 | ||
BC104741.1 | - | 2 | 715 | 40022689 |