Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snap23 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022689.2 | NP_073180.1 | 6 | 442 | ||
GenBank mRNA | AF052596.1 | - | 294 | |||
BC127494.1 | - | 6 | 442 | 8361815 |