Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RGD1560539 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749961.3 | XP_003750009.1 | 1 | 331 |
Gene Symbol: LOC100911527 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003749962.1 | XP_003750010.1 | 1 | 248 |