Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100911180 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_003752173.3 | XP_003752221.2 | 2, 3 | 229 | ||
XM_008773705.1 | XP_008771927.1 | 1, 2 | 793 |