Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SP140 SIRNA Assays ›
Gene Symbol:
SP140
Gene Name:
SP140 nuclear body protein
Gene Aliases:
LYSP100, LYSP100-A, LYSP100-B
Chromosome Location:
Chr.2: 230202924 - 230316571 on Build GRCh38
UniGene ID:
Hs.632549
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22240
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SP140
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001005176.2
NP_001005176.1
1, 2
176
NM_001278451.1
NP_001265380.1
1, 2
176
NM_001278452.1
NP_001265381.1
1, 2
176
NM_001278453.1
NP_001265382.1
1, 2
176
NM_007237.4
NP_009168.4
1, 2
176
XM_005246252.2
XP_005246309.1
1, 2
140
XM_005246253.2
XP_005246310.1
1, 2
140
XM_005246254.2
XP_005246311.1
1, 2
140
XM_005246255.2
XP_005246312.1
1, 2
140
XM_005246256.2
XP_005246313.1
1, 2
140
XM_006712223.3
XP_006712286.1
1, 2
140
XM_011510515.1
XP_011508817.1
1, 2
140
XM_011510516.1
XP_011508818.1
1, 2
140
XM_011510519.1
XP_011508821.1
1, 2
140
XM_011510520.1
XP_011508822.1
1, 2
140
XM_017003239.1
XP_016858728.1
1, 2
140
XM_017003240.1
XP_016858729.1
1, 2
140
XM_017003241.1
XP_016858730.1
1, 2
140
XM_017003242.1
XP_016858731.1
1, 2
140
XM_017003243.1
XP_016858732.1
1, 2
140
XM_017003245.1
XP_016858734.1
1, 2
140
XM_017003246.1
XP_016858735.1
1, 2
140
XM_017003247.1
XP_016858736.1
1, 2
140
XM_017003248.1
XP_016858737.1
1, 2
140
XM_017003249.1
XP_016858738.1
1, 2
140
XM_017003250.1
XP_016858739.1
1, 2
140
XM_017003252.1
XP_016858741.1
1, 2
140
GenBank mRNA
AK301267.1
-
1, 2
176
BC038091.1
-
1, 2
147
BC054890.1
-
1, 2
146
BC070160.1
-
1, 2
142
BC105743.1
-
1, 2
159
40035747
BC105744.1
-
1, 2
161
40035746
BC105960.1
-
1, 2
161
40035752
DC391925.1
-
1, 2
176
U36499.1
-
1, 2
177
U36500.1
-
1, 2
177
U63420.1
-
1, 2
167
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription, DNA-templated
defense response
DNA binding
transcription factor activity, sequence-specific DNA binding
protein binding
zinc ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-7q954:80/10.239.162.42:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0