Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ATXN2L SIRNA Assays ›
Gene Symbol:
ATXN2L
Gene Name:
ataxin 2 like
Gene Aliases:
A2D, A2LG, A2LP, A2RP
Chromosome Location:
Chr.16: 28823048 - 28837237 on Build GRCh38
UniGene ID:
Hs.460499
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s22258
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ATXN2L
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001308230.1
NP_001295159.1
3
584
NM_007245.3
NP_009176.2
3
584
NM_017492.3
NP_059867.3
3
584
NM_145714.2
NP_663760.1
3
584
NM_148414.2
NP_680780.1
3
584
NM_148415.2
NP_680781.1
3
584
NM_148416.2
NP_680782.1
3
584
XM_005255061.1
XP_005255118.1
3
584
XM_005255062.1
XP_005255119.1
3
584
XM_005255063.3
XP_005255120.1
3
584
XM_005255064.1
XP_005255121.1
3
584
XM_005255065.1
XP_005255122.1
3
584
XM_005255066.1
XP_005255123.1
3
584
XM_005255067.1
XP_005255124.1
3
584
XM_005255068.1
XP_005255125.1
3
584
XM_005255069.1
XP_005255126.1
3
584
XM_005255070.1
XP_005255127.1
3
584
XM_005255071.1
XP_005255128.1
3
584
XM_005255074.1
XP_005255131.1
3
584
XM_005255075.1
XP_005255132.1
3
584
XM_005255076.1
XP_005255133.1
3
584
XM_005255077.1
XP_005255134.1
3
467
XM_006721007.1
XP_006721070.1
3
584
XM_006721008.1
XP_006721071.1
3
584
XM_006721009.1
XP_006721072.1
3
584
XM_006721010.2
XP_006721073.1
3
584
XM_006721011.1
XP_006721074.1
3
584
XM_006721012.3
XP_006721075.1
3
584
XM_011545719.2
XP_011544021.1
3
584
XM_011545721.2
XP_011544023.1
3
584
XM_011545722.1
XP_011544024.1
3
284
XM_017022891.1
XP_016878380.1
3
467
XM_017022892.1
XP_016878381.1
3
284
GenBank mRNA
AF034373.1
-
5
686
AJ317970.1
-
3
540
AJ317971.2
-
3
540
AJ317972.2
-
3
540
AJ317973.2
-
3
540
AJ317974.2
-
3
540
AK289981.1
-
3
584
AY188334.1
-
5
686
AY188335.1
-
5
686
AY188336.1
-
5
686
AY188337.1
-
5
686
AY188338.1
-
5
686
BC068012.1
-
4
442
BC082760.1
-
3
380
5812230
BC136584.1
-
3
537
9020574
U70671.1
-
3
214
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of cytoplasmic mRNA processing body assembly
stress granule assembly
cell-cell adhesion
protein binding
poly(A) RNA binding
cadherin binding involved in cell-cell adhesion
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-qqtbl:80/10.239.164.61:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0