Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Aldh6a1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_031057.2 | NP_112319.2 | 6 | 646 | ||
XM_006240354.2 | XP_006240416.1 | 6 | 1011 | |||
XM_008764862.1 | XP_008763084.1 | 5 | 550 | |||
XM_008764863.1 | XP_008763085.1 | 6 | 648 | |||
GenBank mRNA | DN948946.1 | - | 6 | 643 |