Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr376 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001172686.1 | NP_001166157.1 | 3 | 1057 | ||
NM_146922.2 | NP_667133.1 | 2 | 926 |
Gene Symbol: Olfr20 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146923.2 | NP_667134.2 | 2 | 909 | ||
GenBank mRNA | BC138172.1 | - | 1 | 812 | 8861191 | |
BC138173.1 | - | 1 | 812 | 8861192 | ||
X89679.1 | - | 1 | 418 | |||
X89692.1 | - | 1 | 418 |