Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GJC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080383.1 | NP_001073852.1 | 3 | 314 | ||
NM_005497.3 | NP_005488.2 | 3 | 318 | |||
XM_005256920.1 | XP_005256977.1 | 3 | 312 | |||
XM_005256921.1 | XP_005256978.1 | 2 | 140 | |||
GenBank mRNA | AK124339.1 | - | 2 | 552 | ||
BC096213.3 | - | 1 | 44 | 40004800 | ||
BC096214.3 | - | 1 | 44 | 40004801 | ||
BC096215.3 | - | 1 | 44 | 40004803 | ||
BC096216.3 | - | 1 | 44 | 40004809 | ||
CN299428.1 | - | 3 | 332 | |||
DA845933.1 | - | 3 | 314 |