Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP30 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001104546.1 | NP_001098016.1 | 11 | 762 | ||
NM_006413.4 | NP_006404.1 | 11 | 762 | |||
XM_011539120.2 | XP_011537422.1 | 14 | 1315 | |||
XM_011539121.2 | XP_011537423.1 | 14 | 1315 | |||
XM_017015475.1 | XP_016870964.1 | 14 | 1315 | |||
XM_017015476.1 | XP_016870965.1 | 13 | 1285 | |||
XM_017015477.1 | XP_016870966.1 | 12 | 1015 | |||
XM_017015478.1 | XP_016870967.1 | 14 | 1315 | |||
GenBank mRNA | AK225532.1 | - | 11 | 735 | ||
AK312900.1 | - | 11 | 736 | |||
AL833628.1 | - | 11 | 747 | |||
BC006991.1 | - | 12 | 1021 | 3827681 | ||
U77665.1 | - | 11 | 754 |