Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: MAP4K1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042600.2 | NP_001036065.1 | 24 | 1945 | ||
NM_007181.5 | NP_009112.1 | 24 | 1945 | |||
XM_011526403.1 | XP_011524705.1 | 25 | 1963 | |||
XM_011526404.1 | XP_011524706.1 | 25 | 1963 | |||
XM_017026231.1 | XP_016881720.1 | 25 | 1971 | |||
GenBank mRNA | AK303267.1 | - | 13 | 1215 | ||
U66464.1 | - | 24 | 1859 |
Gene Symbol: LOC105372397 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BU738492.1 | - | 2 | 337 |