Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OSBPL10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174060.1 | NP_001167531.1 | 3 | 1191 | ||
NM_017784.4 | NP_060254.2 | 3 | 1191 | |||
XM_005264843.3 | XP_005264900.1 | 3 | 967 | |||
XM_005264844.4 | XP_005264901.1 | 3 | 967 | |||
XM_011533326.2 | XP_011531628.1 | 3 | 344 | |||
XM_017005667.1 | XP_016861156.1 | 3 | 857 | |||
XM_017005668.1 | XP_016861157.1 | 3 | 834 | |||
XM_017005669.1 | XP_016861158.1 | 3 | 363 | |||
GenBank mRNA | AF346291.1 | - | 3 | 637 | ||
AF392451.1 | - | 4 | 901 | |||
AK000370.1 | - | 1 | 67 | |||
AK304067.1 | - | 3 | 644 | |||
AL833247.1 | - | 3 | 294 | |||
BC003168.1 | - | 3 | 595 | 2905981 |