Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGXT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000030.2 | NP_000021.1 | 6, 7 | 812 | ||
GenBank mRNA | AB292648.1 | - | 6, 7 | 713 | ||
AF348451.1 | - | 6, 7 | 603 | |||
AK292754.1 | - | 6, 7 | 733 | |||
AK315331.1 | - | 6, 7 | 760 | |||
BC132819.1 | - | 6, 7 | 809 | 40146841 | ||
D13368.1 | - | 6, 7 | 799 | |||
X53414.1 | - | 6, 7 | 813 | |||
X56092.1 | - | 6, 7 | 712 |