Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXO45 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001105573.1 | NP_001099043.1 | 2 | 743 | ||
GenBank mRNA | AK025697.1 | - | 2 | 471 | ||
BE778263.1 | - | 2 | 392 |