Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TUBB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293212.1 | NP_001280141.1 | 3 | 614 | ||
NM_001293213.1 | NP_001280142.1 | 3 | 530 | |||
NM_001293214.1 | NP_001280143.1 | 2 | 398 | |||
NM_001293215.1 | NP_001280144.1 | 3 | 517 | |||
NM_001293216.1 | NP_001280145.1 | 3 | 416 | |||
NM_178014.3 | NP_821133.1 | 3 | 530 | |||
NR_120608.1 | 3 | 530 | ||||
GenBank mRNA | AB062393.1 | - | 3 | 356 | ||
AF070561.1 | - | 3 | 351 | |||
AF070593.1 | - | 3 | 351 | |||
AF070600.1 | - | 3 | 368 | |||
AF141349.1 | - | 3 | 368 | |||
AK098772.1 | - | 3 | 375 | |||
AK297489.1 | - | 3 | 284 | |||
AK298889.1 | - | 3 | 420 | |||
AK302314.1 | - | 3 | 483 | |||
AK316265.1 | - | 3 | 287 | |||
AK316316.1 | - | 3 | 412 | |||
BC001938.1 | - | 3 | 355 | 2820585 | ||
BC002347.2 | - | 3 | 360 | 2964461 | ||
BC005838.2 | - | 3 | 355 | 2820585 | ||
BC007605.1 | - | 3 | 360 | 3350604 | ||
BC013374.2 | - | 3 | 351 | 3946253 | ||
BC019924.1 | - | 3 | 348 | 4998571 | ||
BC020946.1 | - | 3 | 389 | 4795692 | ||
BC021909.1 | - | 3 | 350 | 3509417 | ||
BC062532.1 | - | 3 | 348 | |||
BC070326.1 | - | 3 | 366 | 6698679 | ||
BC103746.1 | - | 3 | 373 | 6503977 | ||
BE245819.1 | - | 3 | 384 | |||
BU557299.1 | - | 3 | 530 | |||
DC353572.1 | - | 3 | 518 | |||
DC366174.1 | - | 3 | 513 | |||
DC389555.1 | - | 2 | 242 | |||
DC405540.1 | - | 3 | 327 | |||
V00599.1 | - | 2 | 203 |