Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FCGR2A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001136219.1 | NP_001129691.1 | 4 | 471 | ||
NM_021642.3 | NP_067674.2 | 4 | 468 | |||
XM_011509287.2 | XP_011507589.1 | 4 | 456 | |||
XM_011509290.2 | XP_011507592.1 | 4 | 456 | |||
XM_011509291.1 | XP_011507593.1 | 4 | 456 | |||
XM_017000663.1 | XP_016856152.1 | 4 | 453 | |||
XM_017000664.1 | XP_016856153.1 | 4 | 456 | |||
XM_017000665.1 | XP_016856154.1 | 4 | 456 | |||
XM_017000666.1 | XP_016856155.1 | 4 | 456 | |||
XM_017000667.1 | XP_016856156.1 | 4 | 456 | |||
XM_017000668.1 | XP_016856157.1 | 4 | 453 | |||
GenBank mRNA | AK225438.1 | - | 4 | 429 | ||
AK225601.1 | - | 4 | 424 | |||
AK226059.1 | - | 4 | 440 | |||
AK298657.1 | - | 4 | 542 | |||
AK315225.1 | - | 4 | 429 | |||
BC019931.1 | - | 4 | 453 | 4536764 | ||
BC020823.1 | - | 4 | 418 | 4704252 | ||
J03619.1 | - | 4 | 423 | |||
M28697.1 | - | 4 | 405 | |||
M31932.1 | - | 4 | 425 | |||
X62572.1 | - | 4 | 405 | |||
Y00644.1 | - | 4 | 437 |