Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PMPCA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282944.1 | NP_001269873.1 | 12 | 1428 | ||
NM_001282946.1 | NP_001269875.1 | 13 | 1521 | |||
NM_015160.2 | NP_055975.1 | 13 | 1523 | |||
XM_017014543.1 | XP_016870032.1 | 12 | 1384 | |||
GenBank mRNA | AK129769.1 | - | 2 | 1329 | ||
AK296617.1 | - | 12 | 1340 | |||
AK308104.1 | - | 13 | 1436 | |||
BC004342.1 | - | 4 | 282 | |||
BC022949.1 | - | 13 | 1415 | |||
BC033103.1 | - | 13 | 1432 | |||
BC111399.1 | - | 13 | 1438 | 4609491 | ||
BC132724.1 | - | 13 | 1462 | 40146746 | ||
BC136599.1 | - | 13 | 1462 | 9020589 | ||
D21064.1 | - | 13 | 1436 | |||
D50913.2 | - | 13 | 1436 |