Gene Symbol: GCNT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001491.2 | NP_001482.1 | 3 | 1839 | ||
NM_145649.4 | NP_663624.1 | 5 | 1688 | |||
NM_145655.3 | NP_663630.2 | 3 | 1367 | |||
XM_005248999.2 | XP_005249056.1 | 6 | 1486 | |||
XM_006715052.3 | XP_006715115.1 | 5 | 1765 | |||
GenBank mRNA | AB078432.1 | - | 3 | 1134 | ||
AB078433.1 | - | 3 | 1137 | |||
AF401652.1 | - | 3 | 1248 | |||
AF401653.1 | - | 3 | 1248 | |||
AF458024.1 | - | 3 | 1137 | |||
AF458025.1 | - | 3 | 1131 | |||
AF458026.1 | - | 3 | 1137 | |||
AF458027.1 | - | 3 | 1137 | |||
AF458028.1 | - | 3 | 1137 | |||
AF458029.1 | - | 3 | 1137 | |||
AF458030.1 | - | 3 | 1137 | |||
AK090483.1 | - | 3 | 1693 | |||
AK098105.1 | - | 3 | 531 | |||
AK291195.1 | - | 3 | 1423 | |||
AK291767.1 | - | 5 | 1672 | |||
AK307329.1 | - | 4 | 495 | |||
AK307337.1 | - | 5 | 568 | |||
AK307373.1 | - | 5 | 696 | |||
AK313426.1 | - | 3 | 1382 | |||
AK313903.1 | - | 3 | 1548 | |||
AL832714.1 | - | 3 | 390 | |||
AL832719.1 | - | 3 | 1368 | |||
AY435145.1 | - | 3 | 1137 | |||
AY435146.1 | - | 3 | 1131 | |||
AY435147.1 | - | 3 | 1137 | |||
BC074801.2 | - | 3 | 1151 | 30915526 | ||
BC074802.2 | - | 3 | 1151 | 30915312 | ||
BC130524.1 | - | 3 | 1309 | 40146555 | ||
BX647576.1 | - | 3 | 1366 | |||
BX647668.1 | - | 3 | 1368 | |||
L19659.1 | - | 3 | 1385 | |||
Z19550.1 | - | 3 | 1385 |