Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC14L3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257378.1 | NP_001244307.1 | 5 | 339 | ||
NM_001257379.1 | NP_001244308.1 | 5 | 339 | |||
NM_001257382.1 | NP_001244311.1 | 5 | 409 | |||
NM_174975.4 | NP_777635.1 | 4 | 291 | |||
XM_011530128.2 | XP_011528430.1 | 4 | 259 | |||
GenBank mRNA | AK298079.1 | - | 5 | 283 | ||
AY158086.1 | - | 4 | 202 | |||
AY240872.1 | - | 4 | 291 | |||
BC069641.1 | - | 4 | 214 | 7262444 | ||
BC101001.2 | - | 5 | 278 | 40006372 | ||
BC101002.1 | - | 5 | 278 | 40006375 | ||
BC101003.1 | - | 5 | 348 | 40006376 | ||
BC101004.1 | - | 4 | 230 | 40006377 | ||
BU584962.1 | - | 4 | 212 |