Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: ADGRF1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_025048.3 | NP_079324.2 | 3 | 322 | ||
NM_153840.3 | NP_722582.2 | 3 | 322 | |||
XM_011514469.1 | XP_011512771.1 | 1 | 37 | |||
GenBank mRNA | AB045123.1 | - | 3 | 276 | ||
AK026337.1 | - | 3 | 253 | |||
AK225460.1 | - | 3 | 253 | |||
AL698860.1 | - | 3 | 306 | |||
BC029358.1 | - | 2 | 147 | |||
BC113634.1 | - | 2 | 135 | |||
BC113636.1 | - | 2 | 135 | |||
CD685935.1 | - | 3 | 316 | |||
DB224175.1 | - | 3 | 324 |