Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USP21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001014443.2 | NP_001014443.1 | 3 | 710 | ||
NM_001319848.1 | NP_001306777.1 | 2 | 569 | |||
NM_012475.4 | NP_036607.3 | 2 | 569 | |||
XM_011509414.1 | XP_011507716.1 | 2 | 632 | |||
XM_017001036.1 | XP_016856525.1 | 2 | 632 | |||
XM_017001037.1 | XP_016856526.1 | 3 | 686 | |||
XM_017001038.1 | XP_016856527.1 | 3 | 722 | |||
XM_017001039.1 | XP_016856528.1 | 2 | 575 | |||
XM_017001040.1 | XP_016856529.1 | 2 | 632 | |||
GenBank mRNA | AB208899.1 | - | 2 | 541 | ||
AF177758.1 | - | 2 | 420 | |||
AF217979.1 | - | 2 | 496 | |||
AF233442.1 | - | 2 | 506 | |||
AK292319.1 | - | 3 | 655 | |||
AK310326.1 | - | 2 | 602 | |||
BC003130.1 | - | 1 | 69 | |||
BC090946.1 | - | 3 | 658 | 3877298 | ||
BC136291.1 | - | 1 | 357 | 9020278 | ||
BI755746.1 | - | 3 | 723 |