Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UQCRQ | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014402.4 | NP_055217.2 | 3 | 1163 | ||
GenBank mRNA | AK297061.1 | - | 2 | 1600 | ||
BU190580.1 | - | 1 | 656 |