Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPUSD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142547.1 | NP_001136019.1 | 3 | 345 | ||
NM_173659.3 | NP_775930.2 | 4 | 390 | |||
XM_011533627.2 | XP_011531929.1 | 4 | 388 | |||
GenBank mRNA | AK092026.1 | - | 4 | 364 | ||
AK093620.1 | - | 1 | 1568 | |||
AK094587.1 | - | 1 | 2011 | |||
AK291888.1 | - | 4 | 391 | |||
AK296202.1 | - | 4 | 377 | |||
AK299556.1 | - | 4 | 388 | |||
BC032135.2 | - | 4 | 364 | 4649090 | ||
BC062362.1 | - | 3 | 321 | 5454417 | ||
BC065741.1 | - | 4 | 392 | 4091801 |