Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM50B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006134.6 | NP_006125.2 | 4 | 473 | ||
NR_040016.1 | 4 | 473 | ||||
XM_011529746.1 | XP_011528048.1 | 4 | 471 | |||
GenBank mRNA | AF045606.2 | - | 4 | 418 | ||
AF086280.1 | - | 3 | 307 | |||
AK309977.1 | - | 4 | 400 | |||
AK310701.1 | - | 4 | 329 | |||
AK311870.1 | - | 3 | 293 | |||
AY358634.1 | - | 4 | 397 | |||
AY820138.1 | - | 3 | 260 | |||
BC000569.2 | - | 4 | 389 | 3342648 | ||
DA286739.1 | - | 4 | 474 | |||
DQ286229.1 | - | 3 | 260 |