Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KCNIP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034837.2 | NP_001030009.1 | 4 | 775 | ||
NM_001034838.2 | NP_001030010.1 | 3 | 622 | |||
NM_001278339.1 | NP_001265268.1 | 3 | 742 | |||
NM_001278340.1 | NP_001265269.1 | 4 | 904 | |||
NM_014592.3 | NP_055407.1 | 3 | 742 | |||
XM_017009407.1 | XP_016864896.1 | 4 | 746 | |||
GenBank mRNA | AF199597.1 | - | 3 | 205 | ||
AI884645.1 | - | 3 | 508 | |||
AK074390.1 | - | 3 | 338 | |||
AK301775.1 | - | 3 | 397 | |||
AY170821.1 | - | 3 | 418 | |||
AY780424.1 | - | 4 | 580 | |||
BC035032.1 | - | 3 | 747 | |||
BC050375.1 | - | 3 | 509 | 5752207 | ||
DQ148476.1 | - | 3 | 232 | |||
DQ148477.1 | - | 3 | 205 | |||
DQ148478.1 | - | 4 | 238 | |||
DQ148479.1 | - | 3 | 205 |