Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001006638.2 | NP_001006639.1 | 6 | 474 | ||
NM_001163989.1 | NP_001157461.1 | 6 | 623 | |||
NM_001163990.1 | NP_001157462.1 | 5 | 363 | |||
NM_175738.4 | NP_783865.1 | 6 | 867 | |||
XM_005257288.4 | XP_005257345.1 | 6 | 492 | |||
GenBank mRNA | AK054846.1 | - | 6 | 804 | ||
AK098068.1 | - | 6 | 474 | |||
AK127073.1 | - | 6 | 1406 | |||
AK290202.1 | - | 6 | 843 | |||
AK296062.1 | - | 6 | 462 | |||
AK296172.1 | - | 5 | 326 | |||
AK298033.1 | - | 5 | 359 | |||
AK303442.1 | - | 6 | 526 | |||
BC016615.1 | - | 6 | 420 | 4520191 | ||
BC040547.1 | - | 6 | 853 | 5287038 |