Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IFNAR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000874.4 | NP_000865.2 | 8 | 1189 | ||
NM_001289125.1 | NP_001276054.1 | 8 | 1189 | |||
NM_207584.2 | NP_997467.1 | 8 | 1143 | |||
NM_207585.2 | NP_997468.1 | 8 | 1143 | |||
GenBank mRNA | AK293059.1 | - | 7 | 826 | ||
BC002793.2 | - | 8 | 1137 | 3626374 | ||
CR541817.1 | - | 7 | 784 | |||
L41942.1 | - | 8 | 1110 | |||
L41944.1 | - | 8 | 1110 | |||
U29584.1 | - | 8 | 850 | |||
X77722.1 | - | 8 | 1009 | |||
X89772.1 | - | 7 | 784 |