Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145036.1 | NP_001138508.1 | 2 | 354 | ||
NM_007357.2 | NP_031383.1 | 2 | 354 | |||
GenBank mRNA | AK295251.1 | - | 2 | 361 | ||
AK302791.1 | - | 2 | 305 | |||
AK312678.1 | - | 2 | 270 | |||
AL832190.1 | - | 2 | 321 | |||
BC014960.1 | - | 2 | 317 | 4876271 | ||
BC051906.1 | - | 2 | 243 | 4541036 | ||
DB449302.1 | - | 2 | 352 | |||
DC349417.1 | - | 2 | 353 | |||
Z34975.1 | - | 2 | 298 |