Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIF5B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004521.2 | NP_004512.1 | 16 | 2220 | ||
XM_017016224.1 | XP_016871713.1 | 16 | 1826 | |||
GenBank mRNA | AK289413.1 | - | 16 | 2166 | ||
AK299542.1 | - | 591 | ||||
AK307502.1 | - | 17 | 1917 | |||
BC126279.1 | - | 16 | 1973 | 8991995 | ||
BC126281.1 | - | 16 | 1973 | 8991997 | ||
FJ224324.1 | - | 16 | 2063 | |||
X65873.1 | - | 16 | 2063 |