Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: COG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145036.1 | NP_001138508.1 | 4 | 491 | ||
NM_007357.2 | NP_031383.1 | 4 | 491 | |||
GenBank mRNA | AK295251.1 | - | 4 | 498 | ||
AK302791.1 | - | 5 | 675 | |||
AK312678.1 | - | 4 | 407 | |||
BC014960.1 | - | 4 | 454 | 4876271 | ||
BC051906.1 | - | 4 | 380 | 4541036 | ||
DC349417.1 | - | 4 | 490 | |||
Z34975.1 | - | 4 | 435 |