Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFEC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001018058.2 | NP_001018068.1 | 3 | 530 | ||
NM_001244583.1 | NP_001231512.1 | 2 | 226 | |||
NM_012252.3 | NP_036384.1 | 4 | 617 | |||
XM_011515963.1 | XP_011514265.1 | 6 | 804 | |||
XM_011515964.2 | XP_011514266.1 | 6 | 819 | |||
XM_011515965.2 | XP_011514267.1 | 6 | 759 | |||
XM_011515966.2 | XP_011514268.1 | 6 | 868 | |||
XM_011515969.1 | XP_011514271.1 | 6 | 804 | |||
XM_011515970.1 | XP_011514272.1 | 5 | 717 | |||
XM_011515972.2 | XP_011514274.1 | 3 | 333 | |||
XM_017011874.1 | XP_016867363.1 | 5 | 920 | |||
XM_017011875.1 | XP_016867364.1 | 5 | 755 | |||
XM_017011876.1 | XP_016867365.1 | 2 | 192 | |||
XM_017011877.1 | XP_016867366.1 | 4 | 581 | |||
XM_017011878.1 | XP_016867367.1 | 2 | 119 | |||
GenBank mRNA | AK301479.1 | - | 6 | 809 | ||
AK313546.1 | - | 4 | 569 | |||
BC029891.1 | - | 3 | 461 | 5179088 | ||
CR933605.1 | - | 3 | 530 | |||
D43945.1 | - | 4 | 546 |