Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MLH1 SIRNA Assays ›
Gene Symbol:
MLH1
Gene Name:
mutL homolog 1
Gene Aliases:
COCA2, FCC2, HNPCC, HNPCC2, hMLH1
Chromosome Location:
Chr.3: 36993350 - 37050846 on Build GRCh38
UniGene ID:
Hs.195364
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224047
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MLH1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000249.3
NP_000240.1
14, 15
1861
NM_001167617.1
NP_001161089.1
14, 15
1656
NM_001167618.1
NP_001161090.1
14, 15
1656
NM_001167619.1
NP_001161091.1
13, 14
1569
NM_001258271.1
NP_001245200.1
14, 15
1861
NM_001258273.1
NP_001245202.1
13, 14
1770
NM_001258274.1
NP_001245203.1
15, 16
1806
XM_005265161.1
XP_005265218.1
12, 13
1544
XM_005265163.1
XP_005265220.1
14, 15
1661
XM_005265164.1
XP_005265221.1
14, 15
1714
XM_005265166.1
XP_005265223.1
13, 14
1575
XM_011533727.1
XP_011532029.1
12, 13
1421
XM_017006449.1
XP_016861938.1
12, 13
1462
XM_017006450.1
XP_016861939.1
12, 13
1507
XM_017006451.1
XP_016861940.1
12, 13
1466
GenBank mRNA
AB209848.1
-
14, 15
1639
AK295359.1
-
14, 15
1635
AK298324.1
-
14, 15
1607
AK298583.1
-
14, 15
1629
AK302807.1
-
16, 17
1590
AK312609.1
-
14, 15
1677
AK316074.1
-
13, 14
1542
AK316264.1
-
13, 14
1587
AY517558.1
-
14, 15
1661
BC006850.1
-
14, 15
1713
3451538
BX648844.1
-
15, 16
1810
EU188670.1
-
5, 6
757
EU188675.1
-
2, 3
256
U07343.1
-
14, 15
1684
U07418.1
-
14, 15
1704
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA poly(A) tail shortening
resolution of meiotic recombination intermediates
mismatch repair
double-strand break repair via nonhomologous end joining
male meiosis chromosome segregation
synapsis
spermatogenesis
intrinsic apoptotic signaling pathway in response to DNA damage
female meiosis chromosome segregation
somatic hypermutation of immunoglobulin genes
meiotic metaphase I plate congression
meiotic telomere clustering
isotype switching
negative regulation of mitotic recombination
oogenesis
meiotic spindle midzone assembly
chromatin binding
single-stranded DNA binding
protein binding
ATP binding
ATPase activity
guanine/thymine mispair binding
MutSalpha complex binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-k9xzd:80/10.239.163.240:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0