Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: OAZ1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301020.1 | NP_001287949.1 | 4, 5 | 669 | ||
NM_004152.3 | NP_004143.1 | 4, 5 | 675 | |||
GenBank mRNA | AK307221.1 | - | 4, 5 | 641 | ||
AL542477.3 | - | 2, 3 | 246 | |||
BC010063.1 | - | 4, 5 | 625 | |||
BM423450.1 | - | 4, 5 | 624 | |||
D78361.1 | - | 4, 5 | 706 | |||
D87914.1 | - | 4, 5 | 622 | |||
U09202.1 | - | 4, 5 | 607 |