Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other NOX4 SIRNA Assays ›
Gene Symbol:
NOX4
Gene Name:
NADPH oxidase 4
Gene Aliases:
KOX, KOX-1, RENOX
Chromosome Location:
Chr.11: 89324353 - 89589611 on Build GRCh38
UniGene ID:
Hs.371036
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s224160
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: NOX4
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001143836.2
NP_001137308.1
7, 8
869
NM_001143837.1
NP_001137309.1
10, 11
1014
NM_001291926.1
NP_001278855.1
7, 8
865
NM_001291927.1
NP_001278856.1
7, 8
619
NM_001291929.1
NP_001278858.1
6, 7
532
NM_001300995.1
NP_001287924.1
7, 8
592
NM_016931.4
NP_058627.1
7, 8
869
XM_006718849.3
XP_006718912.1
7, 8
797
XM_011542857.2
XP_011541159.1
7, 8
669
XM_017017842.1
XP_016873331.1
1, 2
20
XM_017017843.1
XP_016873332.1
7, 8
797
XM_017017844.1
XP_016873333.1
8, 9
855
XM_017017845.1
XP_016873334.1
4, 5
271
GenBank mRNA
AB041035.1
-
7, 8
789
AF254621.1
-
7, 8
636
AF261943.1
-
7, 8
550
AJ704725.1
-
7, 8
546
AJ704726.1
-
7, 8
550
AJ704727.1
-
7, 8
550
AK057189.1
-
10, 11
1014
AK291830.1
-
7, 8
632
AK298323.1
-
7, 8
563
AK298328.1
-
7, 8
619
AK298357.1
-
6, 7
523
AK298376.1
-
3, 4
310
AK311381.1
-
6, 7
532
AK311389.1
-
8, 9
691
AY288918.1
-
7, 8
550
BC040105.1
-
7, 8
622
5590269
BP376400.1
-
6, 7
532
Back To Top
More Information
Panther Classification:
Molecular Function -
oxidase
Gene Ontology Categories:
Function(s)
Process(es)
response to reactive oxygen species
cell morphogenesis
response to hypoxia
superoxide metabolic process
inflammatory response
cell aging
negative regulation of cell proliferation
gene expression
positive regulation of smooth muscle cell migration
superoxide anion generation
positive regulation of apoptotic process
positive regulation of MAP kinase activity
bone resorption
homocysteine metabolic process
positive regulation of stress fiber assembly
positive regulation of protein kinase B signaling
cardiac muscle cell differentiation
oxidation-reduction process
positive regulation of ERK1 and ERK2 cascade
cellular response to cAMP
cellular response to glucose stimulus
cellular response to gamma radiation
cellular response to transforming growth factor beta stimulus
reactive oxygen species metabolic process
positive regulation of reactive oxygen species metabolic process
positive regulation of DNA biosynthetic process
nucleotide binding
protein binding
electron carrier activity
NAD(P)H oxidase activity
superoxide-generating NADPH oxidase activity
oxygen sensor activity
heme binding
flavin adenine dinucleotide binding
modified amino acid binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0