Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: PAFAH1B3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145939.1 | NP_001139411.1 | 6 | 924 | ||
NM_001145940.1 | NP_001139412.1 | 6 | 685 | |||
NM_002573.3 | NP_002564.1 | 5 | 902 | |||
XM_017026846.1 | XP_016882335.1 | 5 | 715 | |||
XM_017026847.1 | XP_016882336.1 | 6 | 880 | |||
XM_017026848.1 | XP_016882337.1 | 6 | 646 | |||
GenBank mRNA | BC003016.1 | - | 5 | 829 | 2822228 | |
BC007863.2 | - | 5 | 700 | 4301185 | ||
CR407626.1 | - | 5 | 567 | |||
D63391.1 | - | 5 | 680 |