Gene Symbol: STK32B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001306082.1 | NP_001293011.1 | 1, 2 | 404 | ||
NM_018401.2 | NP_060871.1 | 1, 2 | 404 | |||
XM_011513497.1 | XP_011511799.1 | 1, 2 | 598 | |||
GenBank mRNA | AJ250839.1 | - | 1, 2 | 121 | ||
AK292696.1 | - | 1, 2 | 371 | |||
AK313846.1 | - | 1, 2 | 406 | |||
BC038238.1 | - | 1, 2 | 98 | 5744969 | ||
BC069203.1 | - | 1, 2 | 325 |