Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHTKD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018706.6 | NP_061176.3 | 5 | 865 | ||
GenBank mRNA | AK309810.1 | - | 5 | 773 | ||
BC002477.2 | - | 5 | 810 | 3347913 | ||
BC007955.2 | - | 5 | 792 | 4302613 |