Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PTGER1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000955.2 | NP_000946.2 | 2 | 1019 | ||
GenBank mRNA | AY275470.1 | - | 1 | 901 | ||
BC039034.1 | - | 2 | 968 | 5748278 | ||
BC051286.1 | - | 2 | 1019 | 6596880 | ||
L22647.1 | - | 2 | 975 |
Set Membership: |
G-protein coupled receptor |